Product Details
- SNP ID
-
rs6499851
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:56683597 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- ATGATCCAGTCCTTTCCTGCAGGGG[C/T]AGCCCAGAGCTTCCCTAGCCTTCCC
- Phenotype
-
MIM: 156355
MIM: 156359
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MT1IP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs59145355] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MT1IP
- Gene Name
- metallothionein 1I, pseudogene
There are no transcripts associated with this gene.
- Gene
- MT1X
- Gene Name
- metallothionein 1X
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005952.3 |
|
Intron |
|
|
NP_005943.1 |
View Full Product Details