Product Details

SNP ID
rs35316515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:62646441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGATCCCAAAGACGGCAGCTCAG[G/T]GTCCTCCACGGAAGACTTGCTCAGG
Phenotype
MIM: 616016
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PPM1H PubMed Links

Gene Details

Gene
PPM1H
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020700.1 3430 UTR 3 NP_065751.1
XM_011538578.2 3430 UTR 3 XP_011536880.1
XM_011538579.1 3430 UTR 3 XP_011536881.1
XM_017019676.1 3430 Intron XP_016875165.1

View Full Product Details