Product Details

SNP ID
rs11156994
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21483716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAACAAAACAACATACATAAATAC[A/C]TGATTTGTGTACTACTGAGAAAAGT
Phenotype
MIM: 607466 MIM: 614032
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
RAB2B PubMed Links

Gene Details

Gene
RAB2B
Gene Name
RAB2B, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
TOX4
Gene Name
TOX high mobility group box family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303523.1 Intron NP_001290452.1
NM_014828.3 Intron NP_055643.1

View Full Product Details