Product Details

SNP ID
rs413948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177029038 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGGCCATATAACAAGCCCCTTTCA[A/G]CCACACCTTGGTACATGTTAATAGG
Phenotype
MIM: 609433 MIM: 605308
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UIMC1 PubMed Links

Gene Details

Gene
UIMC1
Gene Name
ubiquitin interaction motif containing 1
There are no transcripts associated with this gene.

Gene
ZNF346
Gene Name
zinc finger protein 346
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308213.1 Intron NP_001295142.1
NM_001308214.1 Intron NP_001295143.1
NM_001308215.1 Intron NP_001295144.1
NM_001308216.1 Intron NP_001295145.1
NM_001308218.1 Intron NP_001295147.1
NM_001308219.1 Intron NP_001295148.1
NM_001308221.1 Intron NP_001295150.1
NM_001308223.1 Intron NP_001295152.1
NM_012279.3 Intron NP_036411.1
XM_005265860.2 Intron XP_005265917.1
XM_005265864.3 Intron XP_005265921.1
XM_011534493.1 Intron XP_011532795.1
XM_011534494.1 Intron XP_011532796.1
XM_011534495.2 Intron XP_011532797.1
XM_011534496.2 Intron XP_011532798.1
XM_011534497.2 Intron XP_011532799.1
XM_017009294.1 Intron XP_016864783.1
XM_017009295.1 Intron XP_016864784.1
XM_017009296.1 Intron XP_016864785.1
XM_017009297.1 Intron XP_016864786.1
XM_017009298.1 Intron XP_016864787.1
XM_017009299.1 Intron XP_016864788.1
XM_017009300.1 Intron XP_016864789.1
XM_017009301.1 Intron XP_016864790.1
XM_017009302.1 Intron XP_016864791.1
XM_017009303.1 Intron XP_016864792.1

View Full Product Details