Product Details

SNP ID
rs273654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51168810 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAGCTCCCATGGACACCTGAGCA[C/T]CTGTCTGTAGGGACCAGCCCCACAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928517 PubMed Links

Gene Details

Gene
LOC101928517
Gene Name
uncharacterized LOC101928517
There are no transcripts associated with this gene.

Gene
SIGLEC17P
Gene Name
sialic acid binding Ig like lectin 17, pseudogene
There are no transcripts associated with this gene.

View Full Product Details