Product Details

SNP ID
rs13324891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:190849540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCAACTTCACAGCAACTGGAGGA[C/T]CCTGAGGGAATCTTTATAAAGCTCC
Phenotype
MIM: 614448
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GMNC PubMed Links

Gene Details

Gene
GMNC
Gene Name
geminin coiled-coil domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146686.2 Intron NP_001140158.1

View Full Product Details