Product Details

SNP ID
rs5021612
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:6793100 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATCTTATATATATCTCCCCTACC[C/T]GCAAATCCCAAAACAATAGTCATAG
Phenotype
MIM: 611501
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAMTA1 PubMed Links

Gene Details

Gene
CAMTA1
Gene Name
calmodulin binding transcription activator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195563.1 Intron NP_001182492.1
NM_001242701.1 Intron NP_001229630.1
NM_015215.3 Intron NP_056030.1
XM_011541083.2 Intron XP_011539385.1
XM_011541084.2 Intron XP_011539386.1
XM_011541085.2 Intron XP_011539387.1
XM_011541086.2 Intron XP_011539388.1
XM_011541087.2 Intron XP_011539389.1
XM_011541088.2 Intron XP_011539390.1
XM_011541089.2 Intron XP_011539391.1
XM_011541090.2 Intron XP_011539392.1
XM_011541091.2 Intron XP_011539393.1
XM_011541092.2 Intron XP_011539394.1
XM_017000773.1 Intron XP_016856262.1
XM_017000774.1 Intron XP_016856263.1
XM_017000775.1 Intron XP_016856264.1
XM_017000776.1 Intron XP_016856265.1
XM_017000777.1 Intron XP_016856266.1
XM_017000778.1 Intron XP_016856267.1
XM_017000779.1 Intron XP_016856268.1
XM_017000780.1 Intron XP_016856269.1
XM_017000781.1 Intron XP_016856270.1

View Full Product Details