Product Details

SNP ID
rs11864124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84012797 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAGAGTGGAAGGGAAGGCCCTTG[C/G]AGGACCCGGTGTTACTGGCCCGGGC
Phenotype
MIM: 615585
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC38A8 PubMed Links

Gene Details

Gene
SLC38A8
Gene Name
solute carrier family 38 member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080442.2 Intron NP_001073911.1
XM_017022946.1 Intron XP_016878435.1

View Full Product Details