Product Details

SNP ID
rs2255166
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:81239762 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCCCCGGCTTTCCTTTGCTTTCCCG[C/T]TGGTTCAAGGTTTGAGCGCCTGCCG
Phenotype
MIM: 616525
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf89 PubMed Links

Gene Details

Gene
C17orf89
Gene Name
chromosome 17 open reading frame 89
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001086521.1 Intron NP_001079990.1
Gene
LOC105371925
Gene Name
uncharacterized LOC105371925
There are no transcripts associated with this gene.

Gene
SLC38A10
Gene Name
solute carrier family 38 member 10
There are no transcripts associated with this gene.

Gene
TEPSIN
Gene Name
TEPSIN, adaptor related protein complex 4 accessory protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144679.2 Intron NP_653280.1
XM_005257066.2 Intron XP_005257123.1
XM_005257067.2 Intron XP_005257124.1
XM_006721709.3 Intron XP_006721772.1
XM_006721712.2 Intron XP_006721775.1
XM_011524355.1 Intron XP_011522657.1
XM_011524356.1 Intron XP_011522658.1
XM_011524357.1 Intron XP_011522659.1
XM_011524358.2 Intron XP_011522660.1
XM_017024202.1 Intron XP_016879691.1
XM_017024203.1 Intron XP_016879692.1
XM_017024204.1 Intron XP_016879693.1

View Full Product Details