Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014957.2 | Intron | NP_055772.2 | |||
XM_005250838.4 | Intron | XP_005250895.2 | |||
XM_005250839.2 | Intron | XP_005250896.2 | |||
XM_005250840.4 | Intron | XP_005250897.2 | |||
XM_005250841.3 | Intron | XP_005250898.2 | |||
XM_005250842.4 | Intron | XP_005250899.1 | |||
XM_011516933.1 | Intron | XP_011515235.1 | |||
XM_011516934.2 | Intron | XP_011515236.1 | |||
XM_011516937.1 | Intron | XP_011515239.1 | |||
XM_011516938.2 | Intron | XP_011515240.1 | |||
XM_011516939.2 | Intron | XP_011515241.1 | |||
XM_011516940.2 | Intron | XP_011515242.1 | |||
XM_011516941.2 | Intron | XP_011515243.1 | |||
XM_017013241.1 | Intron | XP_016868730.1 | |||
XM_017013242.1 | Intron | XP_016868731.1 | |||
XM_017013243.1 | Intron | XP_016868732.1 |