Product Details

SNP ID
rs10889427
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:63525047 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCCTGCTGTGGAAGATGGGTGTCA[A/G]CTACCACTCATATCCCGATTCTCAT
Phenotype
MIM: 605494
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EFCAB7 PubMed Links

Gene Details

Gene
EFCAB7
Gene Name
EF-hand calcium binding domain 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032437.3 Intron NP_115813.2
XM_005271274.3 Intron XP_005271331.1
XM_006710976.3 Intron XP_006711039.1
XM_006710977.1 Intron XP_006711040.1
XM_011542301.2 Intron XP_011540603.1
XM_017002548.1 Intron XP_016858037.1
Gene
ITGB3BP
Gene Name
integrin subunit beta 3 binding protein
There are no transcripts associated with this gene.

View Full Product Details