Product Details

SNP ID
rs12891954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22885490 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCATATCACCTCAGAAAGCAAAG[A/G]CCCACACTACAGCCTGTGGCTGTTA
Phenotype
MIM: 609921 MIM: 616955
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRP10 PubMed Links

Gene Details

Gene
LRP10
Gene Name
LDL receptor related protein 10
There are no transcripts associated with this gene.

Gene
REM2
Gene Name
RRAD and GEM like GTPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173527.2 Intron NP_775798.2
XM_005267383.3 Intron XP_005267440.1
XM_011536518.2 Intron XP_011534820.1
XM_017021058.1 Intron XP_016876547.1
XM_017021059.1 Intron XP_016876548.1

View Full Product Details