Product Details
- SNP ID
-
rs1814901
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:193242792 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGAATACAGGGCTTCATGGAAACAT[A/C]ACAGTGTAGGAGGATTATGGATGGA
- Phenotype
-
MIM: 606487
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HRASLS
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75362127] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HRASLS
- Gene Name
- HRAS like suppressor
- Gene
- MGC2889
- Gene Name
- uncharacterized protein MGC2889
There are no transcripts associated with this gene.
View Full Product Details