Product Details

SNP ID
rs1617105
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:43645443 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CATGGGGCCAAGGGAGGTGTGGGCG[A/G]GATCTGAGATGAGTGGAATGGGGGA
Phenotype
MIM: 612648
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAD2L1BP PubMed Links

Gene Details

Gene
MAD2L1BP
Gene Name
MAD2L1 binding protein
There are no transcripts associated with this gene.

Gene
RSPH9
Gene Name
radial spoke head 9 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193341.1 Intron NP_001180270.1
NM_152732.4 Intron NP_689945.2
XM_005248901.3 Intron XP_005248958.1
XM_006715014.1 Intron XP_006715077.1
XM_017010426.1 Intron XP_016865915.1

View Full Product Details