Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001029997.3 | 1972 | UTR 3 | NP_001025168.2 | ||
NM_001145665.1 | 1972 | UTR 3 | NP_001139137.1 | ||
XM_005258849.2 | 1972 | UTR 3 | XP_005258906.1 | ||
XM_005258850.2 | 1972 | UTR 3 | XP_005258907.1 | ||
XM_005258851.3 | 1972 | UTR 3 | XP_005258908.1 | ||
XM_006723183.3 | 1972 | UTR 3 | XP_006723246.1 | ||
XM_006723184.3 | 1972 | UTR 3 | XP_006723247.1 | ||
XM_017026739.1 | 1972 | UTR 3 | XP_016882228.1 | ||
XM_017026740.1 | 1972 | UTR 3 | XP_016882229.1 |