Product Details

SNP ID
rs9462861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42990281 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTATGTCCATGAAAGTTTGGGAG[C/T]GTGCACAGTGGGATGCTCTGGATGG
Phenotype
MIM: 143170 MIM: 601646
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEA1 PubMed Links

Gene Details

Gene
MEA1
Gene Name
male-enhanced antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318942.1 Intron NP_001305871.1
NM_001318943.1 Intron NP_001305872.1
NM_014623.3 Intron NP_055438.1
XM_005249121.3 Intron XP_005249178.1
XM_017010868.1 Intron XP_016866357.1
Gene
PPP2R5D
Gene Name
protein phosphatase 2 regulatory subunit B'delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270476.1 Intron NP_001257405.1
NM_006245.3 Intron NP_006236.1
NM_180976.2 Intron NP_851307.1
NM_180977.2 Intron NP_851308.1

View Full Product Details