Product Details

SNP ID
rs885125
Assay Type
Functionally tested
NCBI dbSNP Submissions
61
Location
Chr.1:46546090 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCCCCTTCATGGAGGTCTGTCGC[A/G]CTGTTGTCAGAGAACTGATCCCTGC
Phenotype
MIM: 611455
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KNCN PubMed Links

Gene Details

Gene
KNCN
Gene Name
kinocilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097611.1 2457 UTR 3 NP_001091080.1
NM_001322255.1 2457 UTR 3 NP_001309184.1
Gene
MKNK1-AS1
Gene Name
MKNK1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details