Product Details

SNP ID
rs290186
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:85684609 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGACTATAAGCCTAACTATCTACA[A/G]TGGGTAATTGTGCTAATTATTTTTC
Phenotype
MIM: 612880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC89 PubMed Links

Gene Details

Gene
CCDC89
Gene Name
coiled-coil domain containing 89
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152723.2 1669 UTR 3 NP_689936.1
Gene
SYTL2
Gene Name
synaptotagmin like 2
There are no transcripts associated with this gene.

View Full Product Details