Product Details

SNP ID
rs583245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82765575 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTCTGGCGGGAAAGTCCTTGGCC[A/G]GCGAGAGGACACTGCGGGTGAGCTG
Phenotype
MIM: 604649
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBCD PubMed Links

Gene Details

Gene
TBCD
Gene Name
tubulin folding cofactor D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005993.4 Intron NP_005984.3
XM_005256396.4 Intron XP_005256453.1
XM_005256399.4 Intron XP_005256456.1
XM_005256404.4 Intron XP_005256461.1
XM_006722290.2 Intron XP_006722353.1
XM_006722291.3 Intron XP_006722354.1
XM_006722292.2 Intron XP_006722355.1
XM_011523589.1 Intron XP_011521891.1
XM_011523590.1 Intron XP_011521892.1
XM_011523591.1 Intron XP_011521893.1
XM_011523593.1 Intron XP_011521895.1
XM_011523594.1 Intron XP_011521896.1
XM_011523595.2 Intron XP_011521897.1
XM_011523597.2 Intron XP_011521899.1
XM_011523598.1 Intron XP_011521900.1
XM_011523599.2 Intron XP_011521901.1
XM_011523600.2 Intron XP_011521902.1
XM_017024987.1 Intron XP_016880476.1
XM_017024988.1 Intron XP_016880477.1
XM_017024989.1 Intron XP_016880478.1
XM_017024990.1 Intron XP_016880479.1

View Full Product Details