Product Details

SNP ID
rs2235057
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:126615117 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CTCGTTGTCATTTGTCTTAACACGG[A/G]GCGCGGATTCTCCGGAGAAGGGGAG
Phenotype
MIM: 602575
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMX1B PubMed Links

Gene Details

Gene
LMX1B
Gene Name
LIM homeobox transcription factor 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174146.1 Intron NP_001167617.1
NM_001174147.1 Intron NP_001167618.1
NM_002316.3 Intron NP_002307.2

View Full Product Details