Product Details

SNP ID
rs351725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109226736 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTGGATTAAAACACTGTGGGGAA[A/G]ACTGAGTGACTGATTAATATGTAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100996634 PubMed Links

Gene Details

Gene
LOC100996634
Gene Name
transmembrane protein FLJ37396
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277339.1 Intron NP_001264268.1
XM_017010153.1 Intron XP_016865642.1
XM_017010154.1 Intron XP_016865643.1

View Full Product Details