Product Details

SNP ID
rs12376616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136009203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAAGGAAACTCCAGAGGGAACT[C/T]AAGTGCAGCTCAACCTCACCTACCT
Phenotype
MIM: 615786
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NACC2 PubMed Links

Gene Details

Gene
NACC2
Gene Name
NACC family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144653.4 4233 UTR 3 NP_653254.1
XM_011518221.2 4233 UTR 3 XP_011516523.1

View Full Product Details