Product Details
- SNP ID
-
rs2086098
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:10373949 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTATTTAAAAAGACGATGTAATGA[C/T]CTCTGTTCTGTAACTGAACAATCGG
- Phenotype
-
MIM: 611817
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KLRC4-KLRK1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114070847] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KLRC4-KLRK1
- Gene Name
- KLRC4-KLRK1 readthrough
- Gene
- KLRK1
- Gene Name
- killer cell lectin like receptor K1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007360.3 |
|
Intron |
|
|
NP_031386.2 |
- Gene
- LOC101928100
- Gene Name
- uncharacterized LOC101928100
There are no transcripts associated with this gene.
View Full Product Details