Product Details

SNP ID
rs36056058
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:32817507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCGGAGGGCGGGGCTGAGGCGC[C/T]TGAGCGGGGCACAACCCTCCTAGAG
Phenotype
MIM: 611889 MIM: 603623
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
S100PBP PubMed Links

Gene Details

Gene
S100PBP
Gene Name
S100P binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256121.1 526 UTR 5 NP_001243050.1
NM_022753.3 526 Intron NP_073590.2
XM_011541961.2 526 Intron XP_011540263.1
XM_011541962.2 526 Intron XP_011540264.1
XM_011541963.1 526 Intron XP_011540265.1
XM_017002098.1 526 Intron XP_016857587.1
Gene
YARS
Gene Name
tyrosyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003680.3 526 UTR 5 NP_003671.1
XM_011542347.1 526 Intron XP_011540649.1
XM_011542348.1 526 Intron XP_011540650.1
XM_017002651.1 526 Intron XP_016858140.1

View Full Product Details