Product Details

SNP ID
rs10512174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:86271659 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCTGGTTCACTTTTTGGACACAA[C/T]GATGATCTTTCATAGTTAAAACAAG
Phenotype
MIM: 611006
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ISCA1 PubMed Links

Gene Details

Gene
ISCA1
Gene Name
iron-sulfur cluster assembly 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030940.3 Intron NP_112202.2

View Full Product Details