Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_013432.4 | Intron | NP_038460.4 | |||
XM_011517048.2 | Intron | XP_011515350.1 | |||
XM_011517049.2 | Intron | XP_011515351.1 | |||
XM_011517050.2 | Intron | XP_011515352.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_016208.3 | Intron | NP_057292.1 | |||
NM_183057.2 | Intron | NP_898880.1 | |||
XM_005272323.3 | Intron | XP_005272380.1 | |||
XM_005272324.3 | Intron | XP_005272381.1 | |||
XM_005272325.3 | Intron | XP_005272382.1 | |||
XM_011517099.1 | Intron | XP_011515401.1 | |||
XM_011517100.1 | Intron | XP_011515402.1 | |||
XM_017013529.1 | Intron | XP_016869018.1 |