Product Details

SNP ID
rs34677431
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:64992207 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AATTCAGCATATACCATATTGCCCA[A/T]GTGTAAAGTAATCCTGCATATCAAA
Phenotype
MIM: 134636
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CHURC1-FNTB PubMed Links

Gene Details

Gene
CHURC1-FNTB
Gene Name
CHURC1-FNTB readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202558.1 Intron NP_001189487.1
NM_001202559.1 Intron NP_001189488.1
Gene
FNTB
Gene Name
farnesyltransferase, CAAX box, beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002028.3 Intron NP_002019.1

View Full Product Details