Product Details

SNP ID
rs12570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6319295 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGCTGGGCTTCCGGCTGGGCTTC[A/T]GCGCTCCGGCCAGGCCCAAGGCGAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PRKCDBP PubMed Links

Gene Details

Gene
PRKCDBP
Gene Name
protein kinase C delta binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145040.2 688 Silent Mutation GCA,GCT A218A NP_659477.2

View Full Product Details