Product Details

SNP ID
rs1149613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:76786388 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACAGAGGAGGTGGCTTTTTGAAT[C/G]AAATGTGTGAGAGTTTGCTGGTAGA
Phenotype
MIM: 608015
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928837 PubMed Links

Gene Details

Gene
LOC101928837
Gene Name
uncharacterized LOC101928837
There are no transcripts associated with this gene.

Gene
TSKU
Gene Name
tsukushi, small leucine rich proteoglycan
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258210.1 Intron NP_001245139.1
NM_001318477.1 Intron NP_001305406.1
NM_001318478.1 Intron NP_001305407.1
NM_001318479.1 Intron NP_001305408.1
NM_015516.3 Intron NP_056331.2

View Full Product Details