Product Details
- SNP ID
-
rs11956870
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:79998554 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAGACATAGCAACACAGACGTAGA[A/C]CCAGACCCAAAATTGGTGAAATCTG
- Phenotype
-
MIM: 600715
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MTX3
PubMed Links
Gene Details
- Gene
- MTX3
- Gene Name
- metaxin 3
There are no transcripts associated with this gene.
- Gene
- THBS4
- Gene Name
- thrombospondin 4
View Full Product Details