Product Details

SNP ID
rs13012455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:148030667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGGAAGGTTGACGAATTCTTTT[C/T]ATTTAATCTTTAGCATTACTTGGAT
Phenotype
MIM: 611472 MIM: 603056
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MBD5 PubMed Links

Gene Details

Gene
MBD5
Gene Name
methyl-CpG binding domain protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018328.4 Intron NP_060798.2
XM_011511470.2 Intron XP_011509772.1
XM_011511472.2 Intron XP_011509774.1
XM_017004482.1 Intron XP_016859971.1
XM_017004483.1 Intron XP_016859972.1
XM_017004484.1 Intron XP_016859973.1
XM_017004485.1 Intron XP_016859974.1
Gene
ORC4
Gene Name
origin recognition complex subunit 4
There are no transcripts associated with this gene.

View Full Product Details