Product Details

SNP ID
rs2862171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12450988 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCACACCCTCACCTGTTTACCCC[C/T]ACGTCCTCAAGACCTGCTTCTTTGT
Phenotype
MIM: 606697
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF443 PubMed Links

Gene Details

Gene
ZNF443
Gene Name
zinc finger protein 443
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005815.4 Intron NP_005806.2
XM_011527622.2 Intron XP_011525924.1
XM_011527623.2 Intron XP_011525925.2
XM_017026139.1 Intron XP_016881628.1
XM_017026140.1 Intron XP_016881629.1

View Full Product Details