Product Details

SNP ID
rs17469542
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:12454882 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTATGGACCATCTGTGTGATTATGG[A/G]GAAAAGTTCAGGACACCAGCAACCT
Phenotype
MIM: 606697
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF443 PubMed Links

Gene Details

Gene
ZNF443
Gene Name
zinc finger protein 443
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005815.4 Intron NP_005806.2
XM_011527622.2 Intron XP_011525924.1
XM_011527623.2 Intron XP_011525925.2
XM_017026139.1 Intron XP_016881628.1
XM_017026140.1 Intron XP_016881629.1
Gene
ZNF709
Gene Name
zinc finger protein 709
There are no transcripts associated with this gene.

View Full Product Details