Product Details

SNP ID
rs2277959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55454979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGGGAGGTGGGGGCGGCCAGG[C/T]GGGCATTACCTCCTTGAACTGGGGG
Phenotype
MIM: 612928
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ISOC2 PubMed Links

Gene Details

Gene
ISOC2
Gene Name
isochorismatase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136201.1 Intron NP_001129673.1
NM_001136202.1 Intron NP_001129674.1
NM_024710.2 Intron NP_078986.1

View Full Product Details