Product Details
- SNP ID
-
rs2112812
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:25236955 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATATAGGTTGGGGTAGTTGGTTGA[A/G]GTGGGCTGTAAGTTTTGTTGAACTT
- Phenotype
-
MIM: 603750
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AQP8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75748493] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AQP8
- Gene Name
- aquaporin 8
There are no transcripts associated with this gene.
- Gene
- ZKSCAN2
- Gene Name
- zinc finger with KRAB and SCAN domains 2
View Full Product Details