Product Details

SNP ID
rs8060972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75573131 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCAAATTCTGAGGGGGAAGTTTA[A/T]CTTCTCAGAGGTTTATGGAATGTGT
Phenotype
MIM: 607452
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GABARAPL2 PubMed Links

Gene Details

Gene
GABARAPL2
Gene Name
GABA type A receptor associated protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007285.6 Intron NP_009216.1

View Full Product Details