Product Details

SNP ID
rs13259546
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:28497696 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGAATGATGGAATGGAGCCCTTTCT[G/T]TAGAGAAAGTGTGAATTGGAATAGA
Phenotype
MIM: 608519 MIM: 606143
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXO16 PubMed Links

Gene Details

Gene
FBXO16
Gene Name
F-box protein 16
There are no transcripts associated with this gene.

Gene
FZD3
Gene Name
frizzled class receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017412.3 Intron NP_059108.1
NM_145866.1 Intron NP_665873.1
XM_017013841.1 Intron XP_016869330.1
XM_017013842.1 Intron XP_016869331.1
XM_017013843.1 Intron XP_016869332.1
XM_017013844.1 Intron XP_016869333.1
Gene
MIR4288
Gene Name
microRNA 4288
There are no transcripts associated with this gene.

View Full Product Details