Product Details

SNP ID
rs10410369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1880558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGGACCTTCCCAGGGGAGCCCA[C/T]GCTGCTGCTGGCAGGGCTATCTCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD17A PubMed Links

Gene Details

Gene
ABHD17A
Gene Name
abhydrolase domain containing 17A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130111.1 Intron NP_001123583.1
NM_031213.3 Intron NP_112490.3
XM_011528338.1 Intron XP_011526640.1
XM_017027341.1 Intron XP_016882830.1
XM_017027342.1 Intron XP_016882831.1

View Full Product Details