Product Details

SNP ID
rs1438389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68055215 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAGACAAGTCCATGTAGAGCCTC[A/G]TAGGTCTTGTTTAGAAGTTTTTGTT
Phenotype
MIM: 610892
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC550113 PubMed Links

Gene Details

Gene
LOC550113
Gene Name
uncharacterized LOC550113
There are no transcripts associated with this gene.

Gene
SYT14P1
Gene Name
synaptotagmin 14 pseudogene 1
There are no transcripts associated with this gene.

Gene
TMPRSS11F
Gene Name
transmembrane protease, serine 11F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207407.2 Intron NP_997290.2
XM_011531935.1 Intron XP_011530237.1

View Full Product Details