Product Details

SNP ID
rs3170216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43204093 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATATGATGTAATGTAATGGTAGA[C/G]GTATGTCATTTGCCCTTTGTACCAA
Phenotype
MIM: 176393
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC284344 PubMed Links

Gene Details

Gene
LOC284344
Gene Name
uncharacterized LOC284344
There are no transcripts associated with this gene.

Gene
PSG4
Gene Name
pregnancy specific beta-1-glycoprotein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276495.1 461 Missense Mutation CTC,GTC L75V NP_001263424.1
NM_001316339.1 461 Intron NP_001303268.1
NM_002780.4 461 Missense Mutation CTC,GTC L75V NP_002771.2
NM_213633.2 461 Missense Mutation CTC,GTC L75V NP_998798.1
XM_017026995.1 461 Intron XP_016882484.1
XM_017026996.1 461 Missense Mutation CTC,GTC L91V XP_016882485.1
XM_017026997.1 461 Missense Mutation CTC,GTC L75V XP_016882486.1
XM_017026998.1 461 Intron XP_016882487.1
XM_017026999.1 461 Intron XP_016882488.1
XM_017027000.1 461 Missense Mutation CTC,GTC L91V XP_016882489.1
XM_017027001.1 461 Intron XP_016882490.1
XM_017027002.1 461 Intron XP_016882491.1

View Full Product Details