Product Details

SNP ID
rs926971
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:39149482 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAGTAGAAGAGGTGGTAAGAAAGAT[A/T]TATCAGATGAGCACTGACCCCAAAC
Phenotype
MIM: 610397
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TRAPPC6B PubMed Links

Gene Details

Gene
TRAPPC6B
Gene Name
trafficking protein particle complex 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079537.1 1680 UTR 3 NP_001073005.1
NM_177452.3 1680 UTR 3 NP_803235.1
XM_011536409.2 1680 UTR 3 XP_011534711.1
XM_017020964.1 1680 Intron XP_016876453.1

View Full Product Details