Product Details
- SNP ID
-
rs6610152
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:38562961 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCCACCACCATATATTAGCCCTA[A/T]TTTTGCTTTGAAACTGGAGCACAGT
- Phenotype
-
MIM: 300096
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TSPAN7
PubMed Links
Gene Details
- Gene
- TSPAN7
- Gene Name
- tetraspanin 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004615.3 |
|
Intron |
|
|
NP_004606.2 |
View Full Product Details