Product Details

SNP ID
rs2060157
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:8950282 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCCTGACTTGGTGTTATATTGTCTC[A/G]AAACAGATTTTGGTCAGGTAGATTA
Phenotype
MIM: 609154
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASCL3 PubMed Links

Gene Details

Gene
ASCL3
Gene Name
achaete-scute family bHLH transcription factor 3
There are no transcripts associated with this gene.

Gene
TMEM9B
Gene Name
TMEM9 domain family member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286094.1 Intron NP_001273023.1
NM_001286095.1 Intron NP_001273024.1
NM_020644.2 Intron NP_065695.1

View Full Product Details