Product Details

SNP ID
rs134981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:26493484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATTTCAATGCTGCGAATCGGCT[A/T]ATGAGCAACACTGGAGCACAGGACT
Phenotype
MIM: 606682 MIM: 602254 MIM: 612747
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HPS4 PubMed Links

Gene Details

Gene
HPS4
Gene Name
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
There are no transcripts associated with this gene.

Gene
SRRD
Gene Name
SRR1 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013694.2 2902 Intron NP_001013716.2
XM_011530178.2 2902 UTR 3 XP_011528480.1
XM_017028799.1 2902 Intron XP_016884288.1
Gene
TFIP11
Gene Name
tuftelin interacting protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008697.1 2902 Intron NP_001008697.1
NM_012143.2 2902 Intron NP_036275.1
XM_011530079.1 2902 Intron XP_011528381.1
XM_017028735.1 2902 Intron XP_016884224.1

View Full Product Details