Product Details

SNP ID
rs739439
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:28396803 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGCCCCTGCACTTACAACTTCCTGC[C/T]GCTCTGTGGCCTTGCCCTGTAATCA
Phenotype
MIM: 607732 MIM: 611672
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SARM1 PubMed Links

Gene Details

Gene
SARM1
Gene Name
sterile alpha and TIR motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015077.3 3063 UTR 3 NP_055892.2
Gene
SLC46A1
Gene Name
solute carrier family 46 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242366.2 3063 UTR 3 NP_001229295.1
NM_080669.5 3063 UTR 3 NP_542400.2
XM_005277786.3 3063 UTR 3 XP_005277843.1
XM_017024110.1 3063 UTR 3 XP_016879599.1

View Full Product Details