Product Details

SNP ID
rs438646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41369284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGGCGGGGCCATTCACACAACC[A/G]TGGTCTTTCCTCCCAGTGAGGAGGG
Phenotype
MIM: 147890
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MX2 PubMed Links

Gene Details

Gene
MX2
Gene Name
MX dynamin like GTPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002463.1 Intron NP_002454.1
XM_005260983.4 Intron XP_005261040.1
XM_005260984.1 Intron XP_005261041.1
XM_011529571.1 Intron XP_011527873.1
XM_011529572.1 Intron XP_011527874.1
XM_011529573.1 Intron XP_011527875.1
XM_011529574.2 Intron XP_011527876.1

View Full Product Details