Product Details
- SNP ID
-
rs722423
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:30599156 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTCAATACGCTATATGAATTTCCA[G/T]GGAAAGGATTGTGCTCTTCTTTACG
- Phenotype
-
MIM: 600287
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GARS
PubMed Links
Gene Details
- Gene
- GARS
- Gene Name
- glycyl-tRNA synthetase
View Full Product Details