Product Details

SNP ID
hCV2271170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:2946040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTACGTTGTCCCCATACAGCC[C/T]GTGGAGACTCTTCCCGAGGAAGTTC
Phenotype
MIM: 300180
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARSE PubMed Links

Gene Details

Gene
ARSE
Gene Name
arylsulfatase E (chondrodysplasia punctata 1)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000047.2 969 Intron NP_000038.2
NM_001282628.1 969 Intron NP_001269557.1
NM_001282631.1 969 Missense Mutation AGG,GGG R272G NP_001269560.1

View Full Product Details