Product Details

SNP ID
rs439926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2890139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACTTGCTGCCTTCATCTTGTGC[C/G]TGCTCAGGGCCCTGCCCTAACCCAG
Phenotype
MIM: 600856 MIM: 602631 MIM: 603240
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDKN1C PubMed Links

Gene Details

Gene
CDKN1C
Gene Name
cyclin dependent kinase inhibitor 1C
There are no transcripts associated with this gene.

Gene
SLC22A18
Gene Name
solute carrier family 22 member 18
There are no transcripts associated with this gene.

Gene
SLC22A18AS
Gene Name
solute carrier family 22 member 18 antisense
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302862.1 658 Intron NP_001289791.1
NM_007105.3 658 Intron NP_009036.2
XM_017017832.1 658 Intron XP_016873321.1
XM_017017833.1 658 Intron XP_016873322.1
XM_017017834.1 658 Intron XP_016873323.1
XM_017017835.1 658 UTR 5 XP_016873324.1
XM_017017836.1 658 Intron XP_016873325.1

View Full Product Details