Product Details

SNP ID
hCV2280232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12892145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCGGGAGATCATTTCGGAGATG[A/G]GGGAGTTGGGTGTGCTGGGCCCCAC
Phenotype
MIM: 608801 MIM: 600599 MIM: 611487
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GCDH PubMed Links

Gene Details

Gene
GCDH
Gene Name
glutaryl-CoA dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000159.3 409 Missense Mutation AGG,GGG R101G NP_000150.1
NM_013976.3 409 Missense Mutation AGG,GGG R101G NP_039663.1
Gene
KLF1
Gene Name
Kruppel like factor 1
There are no transcripts associated with this gene.

Gene
SYCE2
Gene Name
synaptonemal complex central element protein 2
There are no transcripts associated with this gene.

View Full Product Details